Order Entry
Canada
ContactUsLinkComponent
Innovating Science® Distant Learning: Acids and Bases
Innovating Science® Distant Learning: Acids and Bases
Catalog # 470329-730
Supplier:  Ward's Science
undefined
Innovating Science® Distant Learning: Acids and Bases
Catalog # 470329-730
Supplier:  Ward's Science
Supplier Number:  IS1500_U
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Distant learning: acids and bases
  • Cat. no.:
    470329-730
  • No. of Uses:
    2

Specifications

About this item

Learn about acids and bases and what the pH of common household items is.

  • Students will learn how to use pH strips and test common household items to determine if they are acids or bases.
  • Aligned to NGSS Practices
  • Performance expectations: 5-PS1-3, 5-PS1-5
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include cause and effect, scale proportion and quantity

Students will be introduced to acids and bases and test a dilute acid and dilute base with neutral litmus paper to learn how pH strips indicate if something is acidic or basic. Then, they will test common items that could be found around their household to determine if they are acids or bases. Extra litmus paper is included if students want to test more items found in their home to gain a greater understanding of acids and bases. The kit contains an Instruction Manual and enough materials for 2 groups.

Delivery: Kit includes instructional manual and SDS booklet.
Caution: CHOKING HAZARD: Small parts. Not for children under 3 years.