About this item
Designed and purpose-built to meet the demanding needs of scientific, life science, laboratory, and medical cold storage applications.
- Solid and glass door refrigerators from 1 to 15 cubic feet
- Built-in readout for current and high/low temperature
- Audible and visual high/low temperature alarms
- Optimized efficient performance for stable temperature
- Eco-friendly natural refrigerants for a greener footprint
The all-refrigerator units come with an adjustable control range from 2 to 10 °C. The microprocessor temperature controller is externally located to minimize door openings, helping to protect stored contents from ambient temperature. The buffered temperature probe is encased in a glycol-filled bottle to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. The current and min/max temperature display is viewable in Celsius or Fahrenheit to the nearest tenth of a degree. The audible and visual temperature alarm sounds if the unit's interior goes out of range. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. This unit is equipped with remote alarm contacts in the rear and a ½" probe port on the right hand side to accept additional monitoring devices.
Inside, they utilizes adjustable cycle defrost operation to avoid temperature spikes. Optimized forced air cooling with two internal fans ensures excellent temperature stability and uniformity with rapid recovery after door openings. The adjustable plastic-coated wire shelves can be positioned at ½" increments to accommodate virtually any sized item. We include seven shelves, more than competitor products providing to maximize interior shelving storage area. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention.
They come in a white cabinet finish with a blue accented control panel and easy-grip handle. The user-reversible double pane tempered glass door provides a full view of the interior. These models include a factory-installed lock and ships with two keys for staff use. Powerful magnetic gaskets ensure a positive seal.
Designed with hydrocarbon refrigerant, the Pharm-Fac Performance Refrigerators provide an energy efficient solution to vaccine storage. These units are Pyxis, Omnicell, and AcuDose RX compatible.
Specifications
- Catalog No:
- 76636-522
- 76636-524
- 76636-526
- 470328-016
- 470328-000
- 76636-528
- 470327-998
- 470328-004
- 470328-002
- 76636-520
- 76636-496
- 470327-986
- 76636-500
- 470328-006
- 470327-990
- 76636-502
- 470327-988
- 470328-010
- 76636-498
- 470327-994
- 76636-504
- 470328-014
- Model:
- ARS8MLDR
- ARS12MLDR
- ARS15MLDR
- ARS8ML
- ARG8ML
- ARS18ML
- ARG6ML
- ARS15ML
- ARS12ML
- ARS6MLDR
- ARG6MLDR
- ARG12ML
- ARG12MLDR
- ARS1ML
- ARG1ML
- ARG15MLDR
- ARG15ML
- ARS3ML
- ARG8MLDR
- ARG3ML
- ARG18ML
- ARS6ML
- Cat. No.:
- 76636-522
- 76636-524
- 76636-526
- 470328-016
- 470328-000
- 76636-528
- 470327-998
- 470328-004
- 470328-002
- 76636-520
- 76636-496
- 470327-986
- 76636-500
- 470328-006
- 470327-990
- 76636-502
- 470327-988
- 470328-010
- 76636-498
- 470327-994
- 76636-504
- 470328-014
- Alarm:
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Temperature and open door alarms
- Shelves/drawers:
- 6 ventilated drawers
- 6 ventilated drawers
- 6 ventilated drawers
- 7 adjustable wire shelves
- 7 adjustable wire shelves
- 7 adjustable wire shelves
- 4 adjustable wire shelves
- 7 adjustable wire shelves
- 7 adjustable wire shelves
- 3 ventilated drawers
- 3 ventilated drawers
- 7 adjustable wire shelves
- 6 ventilated drawers
- 3 adjustable wire shelves
- 3 adjustable wire shelves
- 6 ventilated drawers
- 7 adjustable wire shelves
- 4 adjustable wire shelves
- 6 ventilated drawers
- 4 adjustable wire shelves
- 7 adjustable wire shelves
- 4 adjustable wire shelves
- Weight:
- 82 kg (180 lbs.)
- 97 kg (214 lbs.)
- 104 kg (230 lbs.)
- 73 kg (160 lbs.)
- 75 kg (166 lbs.)
- 132 kg (290 lbs.)
- 54 kg (119 lbs.)
- 91 kg (200 lbs.)
- 83 kg (184 lbs.)
- 59 kg (130 lbs.)
- 61 kg (134 lbs.)
- 86 kg (190 lbs. )
- 99 kg (219 lbs.)
- 28 kg (61 lbs.)
- 29 kg (63 lbs. )
- 107 kg (235 lbs.)
- 93 kg (205 lbs.)
- 39 kg (85 lbs)
- 84 kg (186 lbs.)
- 40 kg (87 lbs.)
- 136 kg (300 lbs.)
- 52 kg (115 lbs.)
Specifications
Related Information
TDS Medical Laboratory Series Refrigerator with Glass Doors, ARG12ML TDS Medical Laboratory Series Refrigerator with Glass Doors, ARG15ML TDS Medical Laboratory Series Refrigerator with Glass Doors, ARG1ML TDS Medical Laboratory Series Refrigerator with Glass Doors, ARG3ML TDS Medical Laboratory Series Refrigerator with Glass Doors, ARG6ML