Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cap Inserts
VWR® Color Chips for Screw Cap Microtubes
VWR® Color Chips for Screw Cap Microtubes
VWR® Color Chips for Screw Cap Microtubes
Catalog # 76417-202
Supplier:
VWR International
Expand image
VWR® Color Chips for Screw Cap Microtubes
Catalog # 76417-202
Supplier:
VWR International
Specifications
Color:
Assortment
Description:
Color chips
Cat. no.:
76417-202
Specifications
About this item
Product Details
Product Details