VWR® Screw Cap Microtubes with Socket Caps
About this item
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards
- High-clarity design: Crystal-clear walls for easy sample visibility
- Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
- Designed with an insertable ridge for convenient sample coding using colorful insert chips
- Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
- Sterile and non sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
- Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
Ordering information: Spare caps and color insert chips also available.
18 Options Available Below
Product Details & Documents
VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.
Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 & USP Class VI standards
- High-clarity design: Crystal-clear walls for easy sample visibility
- Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
- Designed with an insertable ridge for convenient sample coding using colorful insert chips
- Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
- Sterile and non sterile options: Choose pre-sterilized assemblies or convenient non-sterile combo packs
- Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications
VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.
Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.
Ordering information: Spare caps and color insert chips also available.
Documents
Product Family Options
Screw Cap Microtubes and Socket Caps in Separate Resealable Bags
Product Information
- ColorVolumeCap colorFormatCap typeGraduationsMax. rcfPackagingSterilityAvailabilityPrice
- Clear0.5 mlClearConical bottomInsertable flat capNo20000×gCombo PackNon sterilePack of 500$431.02Case of 5000$4,310.24
- Clear0.5 mlClearFreestandingInsertable flat capNo20000×gCombo PackNon sterilePack of 500$431.02Case of 5000$4,310.24
- Clear1.5 mlClearConical bottomInsertable flat capYes20000×gCombo PackNon sterilePack of 500$415.82Case of 5000$4,158.24
- Clear1.5 mlClearFreestandingInsertable flat capNo20000×gCombo PackNon sterilePack of 500$415.82Case of 5000$4,158.24
- Clear2.0 mlClearConical bottomInsertable flat capYes20000×gCombo PackNon sterilePack of 500$418.11Case of 5000$4,181.12
- Clear2.0 mlClearFreestandingInsertable flat capYes20000×gCombo PackNon sterilePack of 500$394.44Case of 5000$3,944.40
Screw Cap Microtubes with Tethered Socket Caps in Separate Resealable Bags
Product Information
- ColorVolumeCap colorFormatCap typeGraduationsMax. rcfPackagingSterilityAvailabilityPrice
- Clear2.0 mlClearConical baseTethered, socketYes20000×gCombo packNon sterilePack of 500$128.88Case of 5000$1,288.80Specifications:New ProductNew Product
- Clear2.0 mlClearFreestandingTethered, socketYes20000×gCombo packNon sterilePack of 500$112.99Case of 5000$1,129.92Specifications:New ProductNew Product
- Clear0.5 mlClearConical baseTethered, socketNo20000×gCombo packNon sterilePack of 500$128.88Case of 5000$1,288.80Specifications:New ProductNew Product
- Clear0.5 mlClearFreestandingTethered, socketNo20000×gCombo packNon sterilePack of 500$121.18Case of 5000$1,211.76Specifications:New ProductNew Product
- Clear1.5 mlClearConical baseTethered, socketYes20000×gCombo packNon sterilePack of 500$128.88Case of 5000$1,288.80Specifications:New ProductNew Product
- Clear1.5 mlClearFreestandingTethered, socketNo20000×gCombo packNon sterilePack of 500$128.88Case of 5000$1,288.80Specifications:New ProductNew Product
Sterile Assemblies: Screw Cap Microtubes with Attached Socket Caps
Product Information
- ColorVolumeCap colorFormatCap typeGraduationsMax. rcfPackagingSterilityAvailabilityPrice
- Clear0.5 mlClearConical bottomInsertable flat capNo20000×gAssembledSterilePack of 500$547.15Case of 5000$5,471.52
- Clear0.5 mlClearFreestandingInsertable flat capNo20000×gAssembledSterilePack of 500$547.15Case of 5000$5,471.52
- Clear1.5 mlClearConical bottomInsertable flat capYes20000×gAssembledSterilePack of 500$503.92Case of 5000$5,039.17
- Clear1.5 mlClearFreestandingInsertable flat capNo20000×gAssembledSterilePack of 500$535.51Case of 5000$5,355.12
- Clear2.0 mlClearConical bottomInsertable flat capYes20000×gAssembledSterilePack of 500$510.45Case of 5000$5,104.51
- Clear2.0 mlClearFreestandingInsertable flat capYes20000×gAssembledSterilePack of 500$519.68Case of 5000$5,196.80
Customizable Socket Caps
Product Information
- Catalog #DescriptionSterilityCap colorAvailabilityPrice
- Customizable screw capsNon sterileGreenLog in for availabilityPack of 500$224.90Case of 5000$224.90
Color Inserts for Socket Caps
Product Information
- Catalog #ColorDescriptionAvailabilityPrice
Sterile Socket Caps
Product Information
- Catalog #ColorDescriptionAvailabilityPrice