Order Entry
Canada
ContactUsLinkComponent
5-Tetramethylrhodamine-dUTP, 1 mM Solution, Biotium
5-Tetramethylrhodamine-dUTP, 1 mM Solution, Biotium
Catalog # 10018-184
Supplier:  Biotium
CAS Number:  
5-Tetramethylrhodamine-dUTP, 1 mM Solution, Biotium
Catalog # 10018-184
Supplier:  Biotium
Supplier Number:  40001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    5-Tetramethylrhodamine-dUTP, 1mM Solution
  • Size:
    25 µl
  • Storage conditions:
    –20°
  • Cat. no.:
    10018-184
  • Supplier No.:
    40001

Specifications

About this item

5-TAMRA-dUTP (5-Tetramethylrhodamine-dUTP) can be enzymatically incorporated into DNA to make fluorescently labeled probes, and has absorption/emission at 553/577 nm. Note: 5-TAMRA-dUTP may not be compatible with PCR labeling of DNA probes. Supplied as a 1 mM solution in Tris-HCl buffer, pH 7.5.