Specifications
- Flow rate:2 L/min
- Description:Alto I-R (US) Type 1 water system, remote dispense
- Environmentally Preferable:Sustainable Packaging,Sustainable Materials,Low Manufacturing Impact,Energy Efficient,Waste Reducing,Water Efficient
- Model:Alto™
- Inorganics @ 25°C:18.2 MΩ-cm
- Bacteria:<1 CFU/1000 ml
- Endotoxin:<0.001 (with UF Point of Use Filter 76337-284)
- DNases:Free (with UF Point of Use Filter 76337-284)
- Feed water:Purified water (Type II), RO water (Type III)
- RNases:Free (with UF Point of Use Filter 76337-284)
- Resistivity at +25 °C:18.2 MΩ-cm
- TOC content:1 - 5 ppb
- Weight:25 kg
- W×D×H:482.6×508×279.4 mm
- Cat. no.:76337-332
Specifications
About this item
Provides type 1- ultrapure water on demand for applications like cellular analysis, HPLC and mAbs.
- Fast, volumetric and variable dispense, up to 2 L/min
- Real-time TOC and water quality monitoring
- Dual wavelength 185/254 nm UV
- Internal boost pump and leak detector
- Quick changing consumables
- Easy-to-use software and quiet operation
- Under-bench, on-bench, or wall mounted
- Endotoxin/nuclease-free point of use filter available (see 76337-284)
- Feed by pretreated water (RO or DI.)
Alto provides 18.2 M Ω -cm water on demand. The system is fed from a pretreated supply and produces Type 1 water instantly as required. The integrated dispenser or optional remote dispenser provides Type 1 water up to 2 liters per minute. Output is either continuous or volumetrically dispensed. Select the dispense rate required with the intuitive and simple software controls. Alto has quick-change, long-lasting consumables provided in the start up package. Total Organic Carbon (TOC) monitoring is critical when organics will affect results so a real time TOC reading provides absolute confidence in your water. Feedwater quality: <30 µm.
Application: Alto is suitable for Type 1 water usage of up to 120 liters per hour (adequate feed water must be available). Type 1 water is suitable for molecular biology, electrophoresis, cell and tissue culture, sequencing, HPLC, genomics, pharmacology, monoclonal antibodies (mAbs) and any other sensitive applications.