Vernier® Go!Temp
About this item
Go!Temp collect temperature data directly through the USB port of a computer. Essential in any classroom!
We've made it easy to use technology to teach science concepts with Go!Temp. Plug Go!Temp directly into your computer or Chromebook USB port to start collecting real-time temperature data.
Maximum temperature that the sensor can tolerate without damage: 150 °C
Resolution: 0.07 °C
Accuracy: ± 0.5 °C
Response Time: 4 s (to 90% of full reading in water)
Ordering information: Save money when you purchase the Go!Temp Teacher Pack. Each includes 8 Go!Temp USB temperature sensors.
2 Options Available Below
Product Details & Documents
Go!Temp collect temperature data directly through the USB port of a computer. Essential in any classroom!
We've made it easy to use technology to teach science concepts with Go!Temp. Plug Go!Temp directly into your computer or Chromebook USB port to start collecting real-time temperature data.
Maximum temperature that the sensor can tolerate without damage: 150 °C
Resolution: 0.07 °C
Accuracy: ± 0.5 °C
Response Time: 4 s (to 90% of full reading in water)
Ordering information: Save money when you purchase the Go!Temp Teacher Pack. Each includes 8 Go!Temp USB temperature sensors.
Product Family Options
Product Information
- AvailabilityPrice
- CA470018-316Vernier Go! Temp, singleSupplier Number: GO-TEMPSpecifications:New ProductNew Product
- CA470112-954Vernier Go! Temp, teacher packSupplier Number: GT-TPSpecifications:New ProductNew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



