Order Entry
Canada
ContactUsLinkComponent
PIG® Chemotherapy Drug Spill Kits, New Pig
PIG® Chemotherapy Drug Spill Kits, New Pig
Catalog # 76382-744
Supplier:  New Pig
undefined
PIG® Chemotherapy Drug Spill Kits, New Pig
Catalog # 76382-744
Supplier:  New Pig
Supplier Number:  KIT2012

Specifications

  • Description:
    Chemotherapy drug absorbent mat pads
  • Size:
    3 kits/case
  • Cat. no.:
    76382-744
  • Absorbency:
    4 L/kit
  • Details:
    Includes 30 PIG® high-visibility absorbent mat pads, 15W×20"L

Specifications

About this item

Be prepared for hazardous drug spills with a kit that helps meet USP 800 standards.

  • Suitable for safe handling of cytotoxic drugs to minimize the risk of exposure to healthcare workers, patients and the environment
  • Helps to satisfy requirements that spill kits must be part of your chemotherapy spill protocol and readily available in all areas where hazardous drugs are routinely handled
  • Durable 4-mil bag protects kit contents from dirt and moisture; designed for fast response, easy inventory and low-profile storage in a cart or cabinet
  • PIG absorbent mat pads quickly soak up and contain spilled cytotoxins; color-coded yellow to coordinate with chemotherapy materials
  • For use with water-based liquids only

A must-have in places that store, prepare, transport and administer chemotherapy drugs, such as pharmacies, hospitals, healthcare facilities, patient treatment clinics, physician's practice facilities and veterinarian offices.

Ordering information: KIT2010 and KIT2011 includes personal protective equipment (PPE) that meets USP 800 and ASTM standards: includes two pairs of nitrile gloves, one polyethylene-coated gown and one pair of polyethylene-coated boot covers. KIT2012 contains mat pads only.