Order Entry
Canada
ContactUsLinkComponent
 
Strep-Tactin® XT Sepharose Chromatography Resins, Cytiva
 :  Cytiva
 
undefined
Strep-Tactin® XT Sepharose Chromatography Resins, Cytiva
 :  Cytiva

 

  • pH range:
    6 - 10
  • Matrix:
    Rigid, cross-linked agarose, spherical
  • Ligand:
    Strep-Tactin XT
  • Storage temperature:
    2...8 °C in 20% ethanol
  • Binding capacity:
    10 mg protein with Strep-tag II or Twin-Strep-tag/ml of resin
  • Ligand density:
    5 mg/ml resin
  • Flow rate:
    <300 cm/h

 

Frequently Bought Together