Thermo Scientific™ Nalgene™ PC Certified Clean Biotainer Carboys, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
- Sterile
- Clear finish
- Impact resistant polycarbonate construction
These carboys can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0.5 EU/ml endotoxin.
Customers Who Bought This Also Bought