Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Workwear
Kneehigh Stockings
Kneehigh Stockings
Kneehigh Stockings
Catalog # 470093-122
Supplier:
Ward's Science
Expand image
Kneehigh Stockings
Catalog # 470093-122
Supplier:
Ward's Science
Supplier Number:
50620
Specifications
Type:
Knee-high stockings
Garment application:
Casual
,
General Purpose
,
Office
Gender:
For Women
Color:
Assorted
Cat. no.:
470093-122
Supplier No.:
50620
Specifications
About this item
Product Details
Product Details