Pig Recombinant IL2 (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes. IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Pig
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: CTLL-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:P26891
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Interleukin-2|Aldesleukin|TCGF|IL-2|T cell growth factor
- Protein/peptide name:IL2
- Purity:≥95%
- Molecular weight:15.3 kDa
- Sequence:MAPTSSSTKN TKKQLEPLLL DLQLLLKEVK NYENADLSRM LTFKFYMPKQ ATELKHLQCL VEELKALEGV LNLGQSKNSD SANIKESMNN INVTVLELKG SETSFKCEYD DETVTAVEFL NKWITFCQSI YSTLT
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together