Order Entry
Canada
ContactUsLinkComponent
Human Recombinant FGF-5 (Animal Free) (from E. coli)
Human Recombinant FGF-5 (Animal Free) (from E. coli)
New Product
Certificates
undefined
Human Recombinant FGF-5 (Animal Free) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: NR6R-3T3 Proliferation with 1 ug heparin, ED50 ≤10 ng/ml, Bioactivity ≥1.0×10⁵ units/mg
  • Gene ID:
    P12034
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Heparin-binding growth factor 5|HBGF-5|Smag-82
  • Protein/peptide name:
    FGF-5
  • Purity:
    ≥95%
  • Molecular weight:
    27.7 kDa
  • Sequence:
    MAWAHGEKRL APKGQPGPAA TDRNPIGSSS RQSSSSAMSS SSASSSPAAS LGSQGSGLEQ SSFQWSPSGR RTGSLYCRVG IGFHLQIYPD GKVNGSHEAN MLSVLEIFAV SQGIVGIRGV FSNKFLAMSK KGKLHASAKF TDDCKFRERF QENSYNTYAS AIHRTEKTGR EWYVALNKRG KAKRGCSPRV KPQHISTHFL PRFKQSEQPE LSFTVTVPEK KNPPSPIKSK IPLSAPRKNT NSVKYRLKFR FG
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate and 100 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together