Mouse Recombinant IL-10 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Interleukin 10 (IL-10) is an anti-inflammatory cytokine produced by macrophages and type 2 T helper (Th2) cells. IL-10 inhibits the production of pro-inflammatory cytokines such as interferon gamma (IFN-ƴ), tumor necrosis factor alpha (TNF-α), interleukin 2 (IL-2), interleukin 3 (IL-3), interleukin 4 (IL-4), and granulocyte-macrophage colony-stimulating factor (GM-CSF), made by macrophages and regulatory T cells. IL-10 also suppresses antigen presentation on antigen presenting cells, and enhances the survival, proliferation, and antibody production of B cells. Mouse IL-10 is not active on human cells.
- High quality
- Low endotoxin
- Affordable
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity Assay: MC/9 cell proliferation, ED₅₀ ≤5 ng/ml, Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:P18893
- Reconstitution instructions:Sterile water at 0.1 mg/ml (reconstitution at concentrations > 0.1 mg/ml may result in a precipitate)
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Cytokine synthesis inhibitory factor|CSIF
- Protein/peptide name:IL-10 (AF)
- Purity:≥95%
- Molecular weight:18.9 kDa
- Sequence:MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGEKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLQDQGVYKA MNEFDIFINC IEAYMMIKMK S
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together