Corepoint Scientific™ White Diamond Series Refrigerator and Freezer Combination with Controlled Auto Defrost Freezer, Horizon Scientific
These White Diamond combination units provide both refrigeration and freezing capabilities in one convenient unit. These models are designed for scientific and clinical applications.
- Microprocessor temperature control
- Integrated temperature alarms
- Forced draft air circulation
- Probe access
- Keyed door locks
- Controlled Auto Defrost technology
- Natural hydrocarbon refrigerants
These combination units come equipped with forced draft circulation, and precise temperature monitoring through microprocessor control. The freezers on these units feature Controlled Auto Defrost technology which allows for the convenience of auto defrost cycles without high temperature spikes. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
: All units come with a three year parts and labor warranty, plus an additional two year compressor parts warranty.
- Catalog No:
- 76407-440
- 76407-438
- 76407-442
- Cat. No.:
- 76407-440
- 76407-438
- 76407-442
- Defrost:
- Controlled auto defrost freezer/cycle defrost refrigerator
- Controlled auto defrost freezer/cycle defrost refrigerator
- Controlled auto defrost freezer/cycle defrost refrigerator
- Shelves/drawers:
- 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
- 3 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
- 6 adjustable shelves (refrigerator), 2 adjustable shelves (freezer)
- Door type:
- 2 solid swing doors
- 1 glass swing door (refrigerator), 1 solid door (freezer)
- 1 glass swing door (refrigerator), 1 solid door (freezer)
- Shipping weight:
- 143 kg (316 lbs.)
- 94 kg (207 lbs.)
- 131 kg (289 lbs.)
Frequently Bought Together