ContactUsLinkComponent
Infecon® INF-6300 Infectious Substance Shipper, Including COVID-19, Zika, Ebola, Com-Pac
Infecon® INF-6300 Infectious Substance Shipper, Including COVID-19, Zika, Ebola, Com-Pac
Catalog # 76411-526
Supplier:  Com-Pac
undefined
Infecon® INF-6300 Infectious Substance Shipper, Including COVID-19, Zika, Ebola, Com-Pac
Catalog # 76411-526
Supplier:  Com-Pac
Supplier Number:  INF-6300
Specifications
  • Description:
    INF-6300 infecon infectious substance shipper
  • Dimensions:
    15.5×14×12.75"
  • Cat. no.:
    76411-526
Specifications
About this item
The Infecon® 6300 shipper offers the best features of both an overpack shipper and a UN certified category A shipper. The INF-6300 is a combination of both units and provides a shipping solution for any category A or B specimen.
  • Temperature protection for up to 76 hours
  • Holds approximately 22 pounds of dry ice
  • Expanded polystyrene insulated tub
  • Sturdy outer box
  • Infectious substance/dry ice super label
  • Specific instruction sheets for Ebola and Enterovirus shipments
The INF-6300 Shipper is comprised of a 1 liter UN certified vessel, bubble wrap, and a carton with the required UN markings to contain the vessel. This vessel will hold up to 20 standard blood tubes. The vessel (in its carton) is to be placed in the overpack cooler, with refrigerated gel packs or dry ice.
This kit was designed to ship Ebola, Enterovirus, Zika Virus, Measles, and COVID-19 samples. Included in the kit are specific instructions for shipment of Ebola and Enterovirus. It can, however, be used to ship any category A or B specimen.
Ordering information: Components include: Cooler, shipping box, instruction sheets, package return label, dry ice super label, infectious substance dry ice super label, one liter pressure vessel, super infectious substance label (6×7"), shippers declarations, 50ml capacity absorbent sheet (3×3"), perforated six compartment bubble pouch, and foam insert.