ContactUsLinkComponent
Infecon® INF-6200 Infectious Substance Shipper, Com-Pac
Infecon® INF-6200 Infectious Substance Shipper, Com-Pac
Catalog # 76411-524
Supplier:  Com-Pac
undefined
Infecon® INF-6200 Infectious Substance Shipper, Com-Pac
Catalog # 76411-524
Supplier:  Com-Pac
Supplier Number:  INF-6200
Specifications
  • Description:
    INF-6200 infecon infectious substance shipper
  • Dimensions:
    15.5×14×12.75"
  • Cat. no.:
    76411-524
Specifications
About this item
The Infecon® INF-6200 shipper offers the best features of both an overpack shipper and a UN certified category A shipper.
  • Temperature protection for up to 76 hours
  • Holds approximately 22 pounds of dry ice
  • Expanded polystyrene insulated tub
  • Sturdy outer box
  • Infectious substance/dry ice super label
  • Specific instruction sheets for Ebola and Enterovirus shipments included
The INF-6200 is comprised of a 0.5 liter UN certified vessel, bubble wrap, and a carton with the required UN markings to contain the vessel. This vessel will hold up to six standard blood tubes. The vessel (in its carton) is to be placed in the overpack cooler, with refrigerated gel packs or dry ice.
This kit was designed to ship Ebola, Enterovirus, Zika Virus, Measles, and COVID-19 samples. Included in the kit are specific instructions for shipment of Ebola and Enterovirus. It can however, be used to ship any category A or B specimen.
Ordering information: Components include: shipping box, stabilizer sheet, instruction sheets, package return label, dry ice super label, infectious substance dry ice super label, 0.5 liter pressure vessel, biological substance label (2×2"), infectious substance label (2×2"), shippers declarations, 50ml capacity absorbent sheet (3×3"), and perforated six compartment bubble pouch.