HiTrap Columns, Capto AVB, Cytiva
Supplier: Cytiva
Certificates
About this item
HiTrap™ columns packed with Capto™ AVB affinity chromatography resin designed for the purification of adeno-associated virus (AAV). Capto™ AVB is based on a rigid base matrix, delivering excellent pressure-flow properties to AAV production.
- Efficient purification of AAV of several subclasses by affinity chromatography
- Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
- High selectivity and excellent scalability
The possibility to run at higher flow rates and bed heights increases flexibility in process design and allows for an increased productivity.
Specifications
- Catalog No:
- CA76333-718
- CA76333-720
- Max. pressure:
- 5 bar
- 5 bar
- Storage:
- 4 to 8 °C, 20% ethanol
- 4 to 8 °C, 20% ethanol
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together