Order Entry
Canada
ContactUsLinkComponent
Micro Plastic Cells, PerkinElmer
Micro Plastic Cells, PerkinElmer
Catalog # CA97024-690
Supplier:  PerkinElmer
Micro Plastic Cells, PerkinElmer
Catalog # CA97024-690
Supplier:  PerkinElmer
Supplier Number:  L7110262

Specifications

  • Capacity:
    70 - 550 µl
  • Description:
    Micro plastic cells
  • Dimensions:
    12.5W×12.5D×45H mm
  • Cat. no.:
    CA97024-690
  • Pathlength:
    10 mm
  • Supplier No.:
    L7110262

Specifications

About this item

These disposable micro cells have a volume of 70 to 550 ml, and a path length of 10 mm.

  • Cells are individually wrapped

Disposable cells greatly reduce the risk of contamination, removing the need for cleaning reagents and reducing the laboratory costs when compared to quartz or glass cells. Each cell is suitable for any UV/Vis spectrophotometer which accommodates cells with a center height of 15 mm.