Torch Knurled Nut, PerkinElmer
Catalog # CA97012-742
Supplier: PerkinElmer
Specifications
- Description:Torch knurled nut
- Cat. no.:CA97012-742
- Supplier No.:N0776027
Specifications
About this item
This torch knurled nut is designed for use with Avio® 200/500 ICP-OES series instruments.
This knurled nut is a part of the Avio® 200 Torch Body Assembly (N0790138) and Avio® 500 Torch Body Assembly (N0810128).