Counter Tops with Flat Top, Stainless Steel, VCTC Series, Advance Tobaco
Supplier: Advance Tabco
Certificates
About this item
The VCTC series is furnished with 1.5×1" square die embossed No-Drip countertop edge with a 0.5" return on four sides.
- 16 gauge stainless steel type 304
- All TIG welded
- Exposed weld areas polished to match adjacent surfaces
- Entire top mechanically polished to a satin finish
- Countertop edge polished to a mirror finish
- Top is sound deadened
- NSF
Roll formed embossed galvanized hat channels are secured to top by means of structural adhesive and weld studs. Engineered and readily adaptable for installation on standard kitchen cabinets. No backsplash.
Customers Who Bought This Also Bought