Specifications
- Description:Faucet-mount personal eyewash
- Material:Brass, chrome plated
- Cat. no.:CAG06767-42
- Supplier No.:G-06767-42
Specifications
About this item
Installs easily on standard faucets, allowing water pressure to keep the stream flowing.
- Easily convert a standard faucet into an eyewash station
- Flow controlled by water pressure
Adjustable outlets are 7.6 cm (3") apart and angled forward. Chrome-plated brass construction. Inlet connection is 55/64"-27 (F) for faucet.