ContactUsLinkComponent
Lumina™ Cobalt (Co) - Copper (Cu) - Iron (Fe) - Manganese (Mn) - Molybdenum (Mo) Hollow Cathode Lamp, PerkinElmer
Lumina™ Cobalt (Co) - Copper (Cu) - Iron (Fe) - Manganese (Mn) - Molybdenum (Mo) Hollow Cathode Lamp, PerkinElmer
Catalog # CA97013-302
Supplier:  PerkinElmer
undefined
Lumina™ Cobalt (Co) - Copper (Cu) - Iron (Fe) - Manganese (Mn) - Molybdenum (Mo) Hollow Cathode Lamp, PerkinElmer
Catalog # CA97013-302
Supplier:  PerkinElmer
Supplier Number:  N3050215
Specifications
  • Description:
    Lumina™ multi-element hollow cathode lamp Cobalt (Co), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo)
  • Cat. no.:
    CA97013-302
  • Supplier No.:
    N3050215
Specifications
About this item
Series N30502XX Lumina™ 2" (50 mm) diameter multi-element lamps are designed to be used with the PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
  • Multi-element lamps are not designed to be used in a furnace system
  • Lumina™ hollow cathode lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Cobalt (Co), Copper (Cu), Iron (Fe), Manganese (Mn), and Molybdenum (Mo). Every genuine PerkinElmer lamp is designed for use with and tested on spectrometers to assure compatibility and the highest performance. A wide range of single-element and multi-element HCLs are available, and are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), lamps are manufactured with a larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume, and therefore the longer the lamp lifetime.