ContactUsLinkComponent
Eluo Nebulizer Cleaner for Glass Nebulizers, PerkinElmer
Eluo Nebulizer Cleaner for Glass Nebulizers, PerkinElmer
Catalog # 10134-608
Supplier:  PerkinElmer
undefined
Eluo Nebulizer Cleaner for Glass Nebulizers, PerkinElmer
Catalog # 10134-608
Supplier:  PerkinElmer
Supplier Number:  N0777712
Specifications
  • Description:
    Eluo nebulizer cleaner
  • Cat. no.:
    10134-608
  • Supplier No.:
    N0777712
Specifications
About this item
This Eluo Nebulizer cleaner is designed for use with newer OpalMist™, PolyCon, DuraMist™, MEINHARD™, and other glass nebulizers. It is important to regularly clean nebulizers since particle build-up in a capillary and tip causes sample flow to be constricted, thereby reducing nebulizer efficiency and performance.
  • Increase nebulizer efficiency and performance
  • Simple three step cleaning process
  • Cleaner is effective on a wide variety of Perkin Elmer nebulizers
Previously there was no simple way to effectively clean and unblock nebulizers. Now, blocked nebulizers can be safely and easily restored to optimum performance with a revolutionary new cleaning instrument, the Eluo. The Eluo is designed to efficiently deliver a cleanser through the nebulizer capillary to dislodge particle build-up and thoroughly clean the nebulizer. Cleaning a nebulizer is now a simple three step process: Fill the Eluo with Methanol. Insert nebulizer, tip first, into the Eluo. Clean the device by pressing the plunger down. Use the Eluo regularly to maintain nebulizer performance and prolong service life. This version of the Eluo is effective for OpalMist™, PolyCon, DuraMist™, SeaSpray™, and MEINHARD™ nebulizers.