Micro Plastic Cells - 10 mm, PerkinElmer
Catalog #: CA97024-686
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Specifications
- Description:Micro plastic cells - 10 mm
- Cat. no.:CA97024-686
- Pathlength:10 mm
- Supplier No.:L7110260
Specifications
About this item
These disposable micro cells have a volume of 70-550 µl, a pathlength of 10 mm and outside dimensions of 45 mm (H)×12.5 mm (W)×12.5 mm (D).
Disposable cells greatly reduce the risk of contamination, removing the need for cleaning reagents and reducing the laboratory costs as compared to quartz or glass cells. Each cell is suitable for any UV/Vis spectrophotometer which accommodates cells with a center height of 15 mm.
Customers Who Bought This Also Bought