ContactUsLinkComponent
Multi-Element Lumina™ Hollow Cathode Lamp Hollow Cathode Lamp, PerkinElmer
Multi-Element Lumina™ Hollow Cathode Lamp Hollow Cathode Lamp, PerkinElmer
Catalog # CA97013-300
Supplier:  PerkinElmer
undefined
Multi-Element Lumina™ Hollow Cathode Lamp Hollow Cathode Lamp, PerkinElmer
Catalog # CA97013-300
Supplier:  PerkinElmer
Supplier Number:  N3050214
Specifications
  • Description:
    Multi-Element Lumina™ HCL
  • Cat. no.:
    CA97013-300
  • Supplier No.:
    N3050214
Specifications
About this item
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Cobalt (Co), Chromium (Cr), Copper (Cu), Manganese (Mn), and Nickel (Ni). Series N30502XX Lumina™ 2 in. (50 mm) diameter multi-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that multi-element lamps are not designed to be used in a furnace system. Every genuine lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available.
The larger the lamp, the greater the inert gas volume and the longer the lamp lifetime. Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.