Order Entry
Canada
ContactUsLinkComponent
Scorpion and Spider Plastomount Set of 4
Scorpion and Spider Plastomount Set of 4
Catalog # CA470324-432
undefined
Scorpion and Spider Plastomount Set of 4
Catalog # CA470324-432
Supplier Number:  CBS4510

Specifications

  • Width:
    7.75"
  • Height:
    1.25"
  • Length:
    7.75"
  • Cat. no.:
    CA470324-432
  • No. of Parts:
    4

Specifications

About this item

Preserved specimens in resin material.

  • High quality display mount

This kit looks into the difference between Scorpions, Tarantula and Spider.

Delivery: This kit includes an information and fact sheet for easy learning.