Order Entry
Canada
ContactUsLinkComponent
Honeybee Life Cycle Plastomount Set
Honeybee Life Cycle Plastomount Set
Catalog # CA470324-428
undefined
Honeybee Life Cycle Plastomount Set
Catalog # CA470324-428
Supplier Number:  CBS1104

Specifications

  • Width:
    7.75"
  • Height:
    1.25"
  • Length:
    7.75"
  • Cat. no.:
    CA470324-428
  • No. of Parts:
    4

Specifications

About this item

Preserved specimens in resin material.

  • High quality display mount

This kit looks into the Life Cycle of a Honey Bee (Apis cerana) in 4 different blocks.

Delivery: This kit includes an information and fact sheet for easy learning.