Order Entry
Canada
ContactUsLinkComponent
 
Frog Plastomount
Catalog # CA470324-538
undefined
Frog Plastomount
Catalog # CA470324-538
Supplier Number:  T411

Specifications

  • Width:
    3"
  • Height:
    1"
  • Length:
    3"
  • Cat. no.:
    CA470324-538
  • No. of Parts:
    1

Specifications

About this item

Preserved specimens in resin material.

  • High quality display mount

This specimen block has a real frog specimen encased. The specimen is preserved and encased in clear resin material. It can be observed from every side to see details.

Delivery: This product comes with its own black box.
Caution: Each specimen is unique and different and may differ from the picture shown.