Order Entry
Canada
ContactUsLinkComponent
Starfish Plastomount
Starfish Plastomount
Catalog # CA470324-632
undefined
Starfish Plastomount
Catalog # CA470324-632
Supplier Number:  TE40

Specifications

  • Width:
    3"
  • Height:
    1"
  • Length:
    3"
  • Cat. no.:
    CA470324-632
  • No. of Parts:
    1

Specifications

About this item

Preserved specimens in resin material.

  • High quality display mount

This paperweight has a REAL Starfish encased in clear square acrylic.

Each Starfish is unique and different and may differ from the picture shown.

Packaging: The Starfish is packaged in its own black box.