About this item
Make your urogenital diagnosis easier, faster, and more cost effective.
- All-in-one test
- Ready-to-use reagents, 'all-liquid' method
- High sensitivity and specificity
- Fast results for clinicians, available within 24 hours
- Visual colorimetric method, without expert interpretation
Detection, identification, and enumeration of urogenital mycoplasmas in one simple test. MYCOFAST® US has been designed for the detection, identification, and enumeration of Ureaplasma uyrealyticum (U.u.) and Mycoplasma hominis (M.h.) in endocervical, urethral, urinary, and semen specimens.
MYCOFAST® US identifies the growth of U.u. and M.h. after 24 hours of incubation. Our exclusive growth activator, the M.h. Supplement, guarantees fast and accurate results. MYCOFAST® US enables the enumeration of U.u. >103 CCU/ml which is compliant with pathological thresholds recognized for semen and urine samples.