ContactUsLinkComponent
Flame Capillary Tubing Assembly for S10 and AS-9X Autosampler Probes, PerkinElmer
Flame Capillary Tubing Assembly for S10 and AS-9X Autosampler Probes, PerkinElmer
Catalog # CA97012-688
Supplier:  PerkinElmer
undefined
Flame Capillary Tubing Assembly for S10 and AS-9X Autosampler Probes, PerkinElmer
Catalog # CA97012-688
Supplier:  PerkinElmer
Supplier Number:  B3000157
Specifications
  • Tubing int.Ø:
    1.0 mm
  • Description:
    Flame capillary tubing assembly for autosampler probe
  • Length:
    1 m
  • Cat. no.:
    CA97012-688
  • Supplier No.:
    B3000157
Specifications
About this item
Replacement capillary for the PerkinElmer flame autosampler probe
  • One red male screw fitting (1/4-28 UNF threads)
The capillary is compatible with sample probes for the PerkinElmer S10 and legacy AS-9X autosamplers (AS-90, AS-90A, AS-90 Plus, AS-91, and AS-93 Plus) .