2-Port Vent Assembly, Straight, GL 45, Universal, 316L Electro Polished Stainless Steel, Chemglass
Catalog #: 76333-466
Supplier: Chemglass
Specifications
- Description:2-port vent assembly, straight, gl-45, universal, 316l electro polished stainless steel, 4 mm od hose barbs, complete with gasket and cap
- Cat. no.:76333-466
Specifications
About this item
Universal vent port assembly is constructed of 316 L electro polished stainless steel. These port assemblies fit all types of spinner flasks, media bottles and vessels. Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes. Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.
Customers Who Bought This Also Bought