ContactUsLinkComponent
Double Port Adapters, M18, for Bioreactors, Chemglass
Double Port Adapters, M18, for Bioreactors, Chemglass
Catalog # 76333-448
Supplier:  Chemglass
undefined
Double Port Adapters, M18, for Bioreactors, Chemglass
Catalog # 76333-448
Supplier:  Chemglass
Supplier Number:  CLS-1380-57
Specifications
  • Description:
    M-18 threaded adapter with two 4 mm ports for #16 pump tubing
  • Cat. no.:
    76333-448
Specifications
About this item
Adapter fits into the headplate and has two connections that are 4 mm OØ to fit precision tubing size 16. Inside the reactor, the 4 mm tubes are 1.25" long.