Support, Distillation Carts, Dual, 5 to 20 L, Chemglass
Supplier: Chemglass
Certificates
About this item
Distillation receiver carts for use with our 10 L through 100 L reactor systems.
- Glassware can be quickly installed and removed by one person
Heavy Duty support cart can be used for round bottom flasks or cylindrical distillation receivers. Rear of cart incorporates ½" stainless steel lattice for mounting support clamps or accessories.
Frequently Bought Together