Order Entry
Canada
ContactUsLinkComponent
Butterfly Plastomount Set, 4 Specimens
Butterfly Plastomount Set, 4 Specimens
Catalog # CA470324-364
Butterfly Plastomount Set, 4 Specimens
Catalog # CA470324-364
Supplier Number:  BFK1103

Specifications

  • Width:
    7.75"
  • Height:
    1.25"
  • Length:
    7.75"
  • Cat. no.:
    CA470324-364
  • No. of Parts:
    4

Specifications

About this item

Preserved specimens in resin material.

  • High quality display mount

This unique Biology for kids collection introduces children to the  different stages of the butterfly life cycle (Cethosia biblis (Drury)) from the egg to an adult. This kit comes in its own box and comes complete with a Detailed Information Booklet for children ranging from JK to Grade six. Each specimen is unique and different and may differ from the picture shown. Learning can be fun for kids.