GemCone Low-Flow Nebulizer for Optima® 3X00, PerkinElmer
Catalog # CA97013-594
Supplier: PerkinElmer
Specifications
- Description:GemCone low-low nebulizer for Optima® 3X00
- Cat. no.:CA97013-594
- Supplier No.:N0690671
Specifications
About this item
This GemCone nebulizer is designed for use with the Optima® 3000 XL or the Optima® 3000 DV in the axial configuration. Low-flow GemCone Nebulizers permit lower nebulizer gas flow rates, useful for spectral lines with high excitation energies and for providing a more robust plasma.
GemCone Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.