Mini Cuvette Stirrers, FireflySci
Catalog #: 76016-690
Supplier: FIREFLYSCI, INC.
Specifications
- Ø:2 mm
- Length:7 mm
- Description:Mini spectrophotometer precision cell stirrer
- Cat. no.:76016-690
Specifications
About this item
Mini Spectrophotometer Precision Cell Stirrer: This PTFE magnetic centrifugal stirrer fits into cuvettes with smaller apertures and volumes and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine.
- Dimensions: 7×2 mm
- For use with narrow-necked and semi-micro cuvettes
Centrifugal pumping action generated by the cross channels in the upper face mixers without aeration. Photometric measurements can proceed uninterrupted while reagents are added.
Customers Who Bought This Also Bought