NIST 2031 UV/VIS Photometric Accuracy Kit, FireflySci
Catalog # 76016-842
Supplier: FIREFLYSCI, INC.
CAS Number:
Specifications
- Description:NIST 2031 UV/VIS photometric accuracy kit (10%T, 30%T, 90%T)
- Cat. no.:76016-842
- Tolerance:FUV-0.04 (±0.0019 au), FUV-0.5 (±0.0039 au), FUV-1 (±0.0042 au)
Specifications
About this item
Firefly's upgraded and improved version of the NIST-2031 UV/VIS photometric accuracy kit.
- Photometric accuracy in the UV and VIS range
- Stray light performance in the UV and VIS range
- Optical densities: 0.04 au (90%), 0.5 au (30%), 1.0au (10%)
- Wavelength range: 200 to 700 nm (can be used for any wavelength within this range)
- External dimensions of individual filters: 12.5×12.5×45mm
Delivery: This kits includes the following filters: FUV-0.04 (90%T), FUV-0.5 (30%T), FUV-1.0 (10%T), A30 blank holder, and case P4.