Orders LinkContactUsLinkComponent
Toad Dissection Plastomount
Toad Dissection Plastomount
Toad Dissection Plastomount
Certificates
Toad Dissection Plastomount
Toad Dissection Plastomount
2 Options Available Below
Product Details & Documents
Preserved toad dissection in resin material.
  • High quality display mount
This Block looks at a real toad dissection. The specimen is preserved and encased in clear resin material. It can be observed from every side to see details.
The specimen is a toad (Bufo garagarizans). Teacher information guide, student activity sheet, and answer key are included.
Product Family Options
Product Information
  • Height
    Length
    Width
    No. of parts
    Availability
    Price
  • CA470324-568
    Small toad dissection plastomount
    Small toad dissection plastomount
    1.325"
    6"
    3.125"
    1
    Log in for availability
    Log in for availability
    Each
    Each - $215.65
    $215.65
    Each
       $215.65
    Minimum order quantity is 1
    Supplier Number: T604S
  • CA470324-566
    Large toad dissection plastomount
    Large toad dissection plastomount
    1.500"
    7.5"
    4.000"
    1
    Log in for availability
    Log in for availability
    Each
    Each - $381.50
    $381.50
    Each
       $381.50
    Minimum order quantity is 1
    Supplier Number: T604