Avantor® ACE® SuperC18, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® SuperC18™ columns provide high stability across an extended pH range.
- Ultra-inert for maximum performance and reproducibility
- Ultra low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1.7 to 10 µm) and complementary phases
These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
Specifications
- Catalog No:
- 76386-028
- 76386-120
- 76386-072
- 76386-040
- 76386-042
- 76386-074
- 76386-060
- 76386-174
- 76385-998
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- 14.8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- 275 bar (2900.7 psi)
- pH range:
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- 1.5 - 11.5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Frequently Bought Together