Cryogenic labels for cryo vials, tubes, microplates, and boxes are perfect for re-labeling samples stored in liquid nitrogen (−196 °C) and low temperature freezers. Labels will adhere under deep-freeze conditions (−80 °C), eliminating the need to thaw samples before labeling.
- Temperature range: −196 to +110 °C (−321 to +230 °F)
- Labels can adhere to already frozen vials (no thawing required)
- Ideal for long-term storage in cryogenic conditions (−196 °C)
- Printable with all major thermal transfer printer brands
Labeling of frozen vials is challenging due to extremely low temperatures, frost, and moisture accumulated on the surface of these cryogenic containers. These labels provide an ideal solution for bio-banks and bio-repositories that need re-labeling of already frozen samples inside liquid nitrogen tanks and freezers. These cryogenic labels are available in roll format for thermal transfer printing. On vials and tubes, it is recommended that these labels are applied with an overlap of at least 0.25" to ensure proper adherence to the frozen surface.