Avantor® ACE® C4-300, HPLC Columns, 10 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® C4-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
- Designed for peptide and protein separations
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.
Specifications
- Catalog No:
- 76391-328
- 76391-314
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 2.6%
- Endcapped:
- Yes
- Max. pressure:
- 275 bar
- pH range:
- 2 - 8
- Surface area:
- 100 m²/g
- USP code:
- L26
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Related Information
Avantor® ACE® Catalogue Avantor® ACE® HPLC UHPLC Columns Phase Guide
Product Literature
Guard holders and accessories
Product Information
Frequently Bought Together