Avantor® Partisil Avantor® Partisphere, HPLC Columns, PAC 5 and 10 µm
Supplier: Avantor
Certificates
About this item
Polar Amino Cyano (PAC) is a bonded phase with secondary amine groups for good thermal and chemical stability.
- Irregular porous silica
- 5 and 10 μm particle sizes
- Hichrom high efficiency
- Stainless steel column hardware
Fast equilibration enables multiple separation mechanisms, including adsorption, reversed-phase and weak anion-exchange. Ideal for carbohydrate separations.
Frequently Bought Together