VeriSpec® ICP Multi-Element Standard 7, RICCA Chemical Company
Catalog # RCRV010717100N
Supplier: Ricca Chemical
CAS Number:
Specifications
- Matrix:5% HNO₃
- Pack type:Natural LDPE bottle
- Size:100 ml
- Cat. no.:RCRV010717100N
- Supplier No.:RV010717-100N
Specifications
About this item
7 component standard.
2500 ppm: Zn
900 ppm: Cr, Pb
800 ppm: Cu
10 ppm: Cd
8 ppm: Hg
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility