Order Entry
Canada
ContactUsLinkComponent
VeriSpec® ICP Multi-Element Standard 10, RICCA Chemical Company
VeriSpec® ICP Multi-Element Standard 10, RICCA Chemical Company
Catalog # RCRV010683100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® ICP Multi-Element Standard 10, RICCA Chemical Company
Catalog # RCRV010683100N
Supplier:  Ricca Chemical
Supplier Number:  RV010683-100N
CAS Number:  

Specifications

  • Matrix:
    10 % HCl/1% HNO₃
  • Pack type:
    Natural LDPE bottle
  • Description:
    VeriSpec® ICP multi-element standard 10
  • Size:
    100 ml
  • Cat. no.:
    RCRV010683100N
  • Supplier No.:
    RV010683-100N

Specifications

About this item

10 component standard.

10 ppm: Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility