Superspher® RP8, HPLC Columns
: Avantor
A range of 4 µm materials has been marketed under the trade name Superspher®.
- Spherical porous silica
- Base deactivated LiChrospher RP-select B
- Endcapping options for RP-8 and RP-18
Hichrom high efficiency packed columns.
- Catalog No:
- 76402-394
- Carbon load:
- 12.5%
- Endcapped:
- No
- Surface area:
- 350 m²/g
Frequently Bought Together