Avantor® Alltima HP Cyano, HPLC Columns
Supplier: HICHROM LIMITED
Certificates
About this item
Avantor® Alltima HP Cyano HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated high purity silica phases
- Multiple selectivity options
- pH stability from 1 to 10
- Extremely stable, long life and reproducible
These columns are ideal for basic drug analysis. The low column bleed of these columns makes them as favourites for microbore applications.
Column hardware: Stainless steel.
Specifications
- Catalog No:
- 75798-614
- 75798-636
- 75798-616
- 75798-638
- 75798-612
- 75798-634
- 76398-382
- 75798-622
- 75798-618
- 75798-620
- 75830-832
- 75830-834
- 75798-630
- 75798-626
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- USP code:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10