ContactUsLinkComponent
Superspher® RP18-E, HPLC Columns
Superspher® RP18-E, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Superspher® RP18-E, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • 76402-370
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Carbon load:
  • 21.6%
  • Endcapped:
  • Yes
  • Surface area:
  • 350 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)