Order Entry
Canada
ContactUsLinkComponent
VeriSpec® ICP Multi-Element Calibration Standard 2A, RICCA Chemical Company
VeriSpec® ICP Multi-Element Calibration Standard 2A, RICCA Chemical Company
Catalog # RCRV010654100N
Supplier:  Ricca Chemical
CAS Number:  
undefined
VeriSpec® ICP Multi-Element Calibration Standard 2A, RICCA Chemical Company
Catalog # RCRV010654100N
Supplier:  Ricca Chemical
Supplier Number:  RV010654-100N
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Natural LDPE bottle
  • Description:
    VeriSpec® ICP multi-element calibration standard 2A
  • Size:
    100 ml
  • Cat. no.:
    RCRV010654100N
  • Supplier No.:
    RV010654-100N

Specifications

About this item

28-components standard in 5% nitric acid.

10 ppm: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, Hg, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Ti, U, V, Zn.

Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility